1-Phenylbut-3-yn-1-amine hydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
(1-Aminobut-3-yn-1-yl)benzene hydrochloride
048270-500MG
048270-1G
101883-552EA
261.54
USD
101883-552
101883-554
1-Phenylbut-3-yn-1-amine hydrochloride ≥95%
1-Phenylbut-3-yn-1-amine hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₂ClN MW: 181.67 g/mol |
MDL Number:
MFCD09865035 CAS Number: 1177093-09-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...