Methyl-2-(1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)acetate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl phthalimid-1-ylacetate

048268-500MG 048268-1G
101883-548EA 278.46 USD
101883-548 101883-550
Methyl-2-(1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)acetate ≥95%
Methyl-2-(1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)acetate

Hazard: Irritant

Formula: C₁₁H₉NO₄
MDL Number: MFCD00158668
CAS Number: 23244-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR