2-[3-(2-Aminophenyl)-3-oxopropyl]-1H-isoindole-1,3(2H)-dione ≥95%

Supplier: Matrix Scientific

Synonyms: N-[3-(2-Aminophenyl)-3-oxopropyl]phthalimide

048267-500MG 048267-1G
101883-544EA 556.92 USD
101883-544 101883-546
2-[3-(2-Aminophenyl)-3-oxopropyl]-1H-isoindole-1,3(2H)-dione ≥95%
2-[3-(2-Aminophenyl)-3-oxopropyl]-1H-isoindole-1,3(2H)-dione

Hazard: Irritant

Formula: C₁₇H₁₄N₂O₃
MW: 294.31 g/mol
MDL Number: MFCD09865034
CAS Number: 17515-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR