2-Iodo-3-nitrotoluene ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Iodo-1-methyl-3-nitrobenzene

048265-500MG 048265-1G 048265-5G
101883-534EA 278.46 USD
101883-534 101883-536 101883-538
2-Iodo-3-nitrotoluene ≥95%
2-Iodo-3-nitrotoluene

Hazard: Irritant

Formula: C₇H₆INO₂
MDL Number: MFCD00091040
CAS Number: 6277-17-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR