Methyl-2-cyano-3-(4-methoxyphenyl)acrylate ≥95%

Supplier: Matrix Scientific

048261-500MG 048261-1G 048261-5G
101883-512EA 181.54 USD
101883-512 101883-514 101883-516
Methyl-2-cyano-3-(4-methoxyphenyl)acrylate ≥95%
Methyl-2-cyano-3-(4-methoxyphenyl)acrylate

Hazard: Irritant

Formula: C₁₂H₁₁NO₃
MDL Number: MFCD00630908
CAS Number: 65576-66-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR