Methyl N-(2,2,4-trimethyl-3-oxo-1-phenylpentyl)carbamate ≥95%

Supplier: Matrix Scientific

048260-500MG 048260-1G
101883-508EA 278.46 USD
101883-508 101883-510
Methyl N-(2,2,4-trimethyl-3-oxo-1-phenylpentyl)carbamate ≥95%
Methyl N-(2,2,4-trimethyl-3-oxo-1-phenylpentyl)carbamate

Hazard: Irritant

Formula: C₁₆H₂₃NO₃
MW: 277.37 g/mol
MDL Number: MFCD09864844
CAS Number: 1000018-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR