2-(4-Cyclohexylphenyl)butanoic acid ≥95%

Supplier: Matrix Scientific

048259-500MG 048259-1G
101883-504EA 747.69 USD
101883-504 101883-506
2-(4-Cyclohexylphenyl)butanoic acid ≥95%
2-(4-Cyclohexylphenyl)butanoic acid

Hazard: Irritant

Formula: C₁₆H₂₂O₂
MW: 246.35 g/mol
MDL Number: MFCD09865032
CAS Number: 41641-69-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR