1-(3,5-Dimethoxyphenyl)-2-(isopropylamino)-1-ethanol ≥95%

Supplier: Matrix Scientific

048258-500MG 048258-1G 048258-5G
101883-498EA 556.92 USD
101883-498 101883-500 101883-502
1-(3,5-Dimethoxyphenyl)-2-(isopropylamino)-1-ethanol ≥95%

Hazard: Irritant

Formula: C₁₃H₂₁NO₃
MDL Number: MFCD09865031
CAS Number: 804428-42-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR