3-[(4-Hydroxyphenyl)sulfinyl]-2,2-dimethylpropanoic acid ≥95%
Supplier: Matrix Scientific
048250-500MG
048250-1G
101883-474EA
1109.23
USD
101883-474
101883-476
3-[(4-Hydroxyphenyl)sulfinyl]-2,2-dimethylpropanoic acid ≥95%
3-[(4-Hydroxyphenyl)sulfinyl]-2,2-dimethylpropanoic acid
Hazard: Irritant
Formula:
C₁₁H₁₄O₄S |
MDL Number:
MFCD09864839 CAS Number: 1171935-44-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...