3-(3-Bromo-4,5-dimethoxyphenyl)acrylic acid ≥95%

Supplier: Matrix Scientific

048248-500MG 048248-1G
101883-468EA 181.54 USD
101883-468 101883-470
3-(3-Bromo-4,5-dimethoxyphenyl)acrylic acid ≥95%
3-(3-Bromo-4,5-dimethoxyphenyl)acrylic acid

Hazard: Irritant

Formula: C₁₁H₁₁BrO₄
MDL Number: MFCD01312552
CAS Number: 51314-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR