Methyl-3-(4-cyanophenyl)-2-oxiranecarboxylate ≥95%

Supplier: Matrix Scientific

048247-500MG 048247-1G
101883-464EA 556.92 USD
101883-464 101883-466
Methyl-3-(4-cyanophenyl)-2-oxiranecarboxylate ≥95%
Methyl-3-(4-cyanophenyl)-2-oxiranecarboxylate

Hazard: Irritant

Formula: C₁₁H₉NO₃
MW: 203.2 g/mol
MDL Number: MFCD09864842
CAS Number: 108492-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR