Methyl-3-(4-nitrophenyl)-2-oxiranecarboxylate ≥95%

Supplier: Matrix Scientific

048246-500MG 048246-1G
101883-460EA 356.92 USD
101883-460 101883-462
Methyl-3-(4-nitrophenyl)-2-oxiranecarboxylate ≥95%
Methyl-3-(4-nitrophenyl)-2-oxiranecarboxylate

Hazard: Irritant

Formula: C₁₀H₉NO₅
MW: 223.19 g/mol
MDL Number: MFCD09865029
CAS Number: 108492-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR