1-(2-Furyl)-2,2-dimethyl-1,3-propanediol ≥95%
Supplier: Matrix Scientific
Synonyms:
2-(1,3-Dihydroxy-2,2-dimethylprop-1-yl)furan
048244-500MG
048244-1G
101883-452EA
921.54
USD
101883-452
101883-454
1-(2-Furyl)-2,2-dimethyl-1,3-propanediol ≥95%
1-(2-Furyl)-2,2-dimethyl-1,3-propanediol
Hazard: Irritant
Formula:
C₉H₁₄O₃ MW: 170.21 g/mol |
MDL Number:
MFCD00720174 CAS Number: 4946-64-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...