(6-Methyl-3-pyridinyl)methanol ≥95%
Supplier: Matrix Scientific
Synonyms:
6-Methyl-3-pyridinemethanol, 5-Hydroxymethyl-2-methylpyridine, (6-Methylpyridin-3-yl)methanol, 5-Hydroxymethyl-2-picoline
048241-500MG
048241-1G
101883-438EA
375.38
USD
101883-438
101883-440
(6-Methyl-3-pyridinyl)methanol ≥95%
(6-Methyl-3-pyridinyl)methanol
Hazard: Irritant
Formula:
C₇H₉NO MW: 123.16 g/mol |
MDL Number:
MFCD08692003 CAS Number: 34107-46-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...