2-(4-Methoxyphenoxy)-1-ethanol ≥95%

Supplier: Matrix Scientific

048240-500MG 048240-1G 048240-5G
101883-432EA 172.31 USD
101883-432 101883-434 101883-436
2-(4-Methoxyphenoxy)-1-ethanol ≥95%
2-(4-Methoxyphenoxy)-1-ethanol

Hazard: Irritant

Formula: C₉H₁₂O₃
MDL Number: MFCD00016568
CAS Number: 5394-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR