3-Ethoxy-2-pyridinamine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Ethoxypyridin-2-amine

048238-500MG 048238-1G
101883-426EA 240 USD
101883-426 101883-428
3-Ethoxy-2-pyridinamine ≥95%
3-Ethoxy-2-pyridinamine

Hazard: Irritant

Formula: C₇H₁₀N₂O
MW: 138.17 g/mol
MDL Number: MFCD00234961
CAS Number: 10006-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR