2-(4-Bromophenyl)butanoic acid ≥95%

Supplier: Matrix Scientific

048237-500MG 048237-1G
101883-422EA 356.92 USD
101883-422 101883-424
2-(4-Bromophenyl)butanoic acid ≥95%
2-(4-Bromophenyl)butanoic acid

Hazard: Irritant

Formula: C₁₀H₁₁BrO₂
MDL Number: MFCD01737245
CAS Number: 99070-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR