2-Hydroxy-2-(4-methylphenyl)acetic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Methyl-D-mandelic acid

048235-1G 048235-5G
101883-412EA 416.92 USD
101883-412 101883-414
2-Hydroxy-2-(4-methylphenyl)acetic acid ≥95%
2-Hydroxy-2-(4-methylphenyl)acetic acid

Hazard: Irritant

Formula: C₉H₁₀O₃
MDL Number: MFCD00961860
CAS Number: 31284-89-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR