Diethyl-2-(1-nitroethyl)succinate ≥95%

Supplier: Matrix Scientific

048232-500MG 048232-1G 048232-5G
101883-396EA 181.54 USD
101883-396 101883-398 101883-400
Diethyl-2-(1-nitroethyl)succinate ≥95%
Diethyl-2-(1-nitroethyl)succinate

Hazard: Irritant

Formula: C₁₀H₁₇NO₆
MDL Number: MFCD11841060
CAS Number: 4753-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR