(4-Chloro-1-benzothiophen-2-yl)methanol ≥95%

Supplier: Matrix Scientific

048229-500MG 048229-1G
101883-384EA 278.46 USD
101883-384 101883-386
(4-Chloro-1-benzothiophen-2-yl)methanol ≥95%
(4-Chloro-1-benzothiophen-2-yl)methanol

Hazard: Irritant

Formula: C₉H₇ClOS
MW: 198.67 g/mol
MDL Number: MFCD11841059
CAS Number: 216575-42-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR