Methyl-5-cyano-2-hydroxybenzenecarboxylate ≥95%

Supplier: Matrix Scientific

048228-500MG 048228-1G
101883-380EA 176.92 USD
101883-380 101883-382
Methyl-5-cyano-2-hydroxybenzenecarboxylate ≥95%
Methyl-5-cyano-2-hydroxybenzenecarboxylate

Hazard: Irritant

Formula: C₉H₇NO₃
MDL Number: MFCD00800949
CAS Number: 84437-12-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR