Ethyl-2,6-dichloro-5-fluoronicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2,6-dichloro-5-fluoropyridine-3-carboxylate

048226-500MG 048226-1G 048226-5G
101883-368EA 181.54 USD
101883-368 101883-370 101883-372
Ethyl-2,6-dichloro-5-fluoronicotinate ≥95%
Ethyl-2,6-dichloro-5-fluoronicotinate

Hazard: Irritant

Formula: C₈H₆Cl₂FNO₂
MW: 238.05 g/mol
MDL Number: MFCD09757511
CAS Number: 82671-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR