3-Bromo-4-[(3,5-dimethylbenzyl)oxy]benzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

048217-500MG 048217-1G 048217-5G
101883-334EA 181.54 USD
101883-334 101883-336 101883-338
3-Bromo-4-[(3,5-dimethylbenzyl)oxy]benzenecarbaldehyde ≥95%
3-Bromo-4-[(3,5-dimethylbenzyl)oxy]benzenecarbaldehyde

Hazard: Irritant

Formula: C₁₆H₁₅BrO₂
MDL Number: MFCD11841052
CAS Number: 1135283-80-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR