(2-Methoxypyridin-4-yl)methanol ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Methoxy-4-pyridinemethanol

048215-500MG 048215-1G
101883-326EA 92.31 USD
101883-326 101883-328
(2-Methoxypyridin-4-yl)methanol ≥95%
(2-Methoxypyridin-4-yl)methanol

Hazard: Irritant

Formula: C₇H₉NO₂
MW: 139.1519 g/mol
MDL Number: MFCD00859298
CAS Number: 123148-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR