Ethyl-4-(difluoromethoxy)benzenecarboxylate ≥95%

Supplier: Matrix Scientific

048194-500MG 048194-1G
101883-252EA 172.31 USD
101883-252 101883-254
Ethyl-4-(difluoromethoxy)benzenecarboxylate ≥95%
Ethyl-4-(difluoromethoxy)benzenecarboxylate

Hazard: Irritant

Formula: C₁₀H₁₀F₂O₃
MDL Number: MFCD06204380
CAS Number: 773134-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR