(5-Nitro-2-piperidinophenyl)methanol ≥95%
Supplier: Matrix Scientific
048189-500MG
048189-1G
048189-5G
101883-234EA
375.38
USD
101883-234
101883-236
101883-238
(5-Nitro-2-piperidinophenyl)methanol ≥95%
(5-Nitro-2-piperidinophenyl)methanol
Hazard: Irritant
Formula:
C₁₂H₁₆N₂O₃ |
MDL Number:
MFCD01312996 CAS Number: 203509-89-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...