2,4,6-Trichlorophenyl-1-butanesulfonate ≥95%

Supplier: Matrix Scientific

048139-500MG 048139-1G 048139-5G
101883-004EA 278.46 USD
101883-004 101883-006 101883-008
2,4,6-Trichlorophenyl-1-butanesulfonate ≥95%
2,4,6-Trichlorophenyl 1-butanesulphonate

Hazard: Irritant

Formula: C₁₀H₁₁Cl₃O₃S
MDL Number: MFCD12498734
CAS Number: 1171919-49-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR