1-(2-Chloroethoxy)-4-nitrobenzene ≥95%

Supplier: Matrix Scientific

048122-1G 048122-5G
101882-904EA 81.54 USD
101882-904 101882-906
1-(2-Chloroethoxy)-4-nitrobenzene ≥95%
1-(2-Chloroethoxy)-4-nitrobenzene

Hazard: Irritant

Formula: C₈H₈ClNO₃
MDL Number: MFCD00674505
CAS Number: 3383-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR