2-Methoxyethyl-2-{5-[2-chloro-4-(trifluoromethyl)phenoxy]-2-nitrophenoxy}propanoate ≥95%

Supplier: Matrix Scientific

048121-500MG 048121-1G 048121-5G
101882-898EA 556.92 USD
101882-898 101882-900 101882-902
2-Methoxyethyl-2-{5-[2-chloro-4-(trifluoromethyl)phenoxy]-2-nitrophenoxy}propanoate ≥95%
2-Methoxyethyl-2-{5-[2-chloro-4-(trifluoromethyl)phenoxy]-2-nitrophenoxy}propanoate

Hazard: Irritant

Formula: C₁₉H₁₇ClF₃NO₇
MDL Number: MFCD02102176
CAS Number: 72082-45-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR