2-(4-Aminophenoxy)acetic acid hydrate ≥95%

Supplier: Matrix Scientific

048107-500MG 048107-1G 048107-5G
101882-826EA 556.92 USD
101882-826 101882-828 101882-830
2-(4-Aminophenoxy)acetic acid hydrate ≥95%
2-(4-Aminophenoxy)acetic acid hydrate

Hazard: Irritant

Formula: C₈H₁₁NO₄
MDL Number: MFCD08056702
CAS Number: 2298-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR