Methyl-5-bromo-2-{[(propylamino)carbonyl]amino}benzenecarboxylate ≥95%
Supplier: Matrix Scientific
048099-500MG
048099-1G
048099-5G
101882-790EA
278.46
USD
101882-790
101882-792
101882-794
Methyl-5-bromo-2-{[(propylamino)carbonyl]amino}benzenecarboxylate ≥95%
Methyl-5-bromo-2-{[(propylamino)carbonyl]amino}benzenecarboxylate
Hazard: Irritant
Formula:
C₁₂H₁₅BrN₂O₃ |
MDL Number:
MFCD11841035 CAS Number: 1135283-68-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...