6-Cyclopropyl-2-oxo-1,2-dihydro-3-pyridine carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Cyclopropyl-2-oxo-1,2-dihydropyridine-3-carboxylic acid

048098-500MG 048098-1G
101882-786EA 556.92 USD
101882-786 101882-788
6-Cyclopropyl-2-oxo-1,2-dihydro-3-pyridine carboxylic acid ≥95%
6-Cyclopropyl-2-oxo-1,2-dihydro-3-pyridine carboxylic acid

Hazard: Irritant

Formula: C₉H₉NO₃
MW: 179.18 g/mol
MDL Number: MFCD11841034
CAS Number: 1135283-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR