[2-(4-Methylpiperazino)-5-nitrophenyl]methanol ≥95%

Supplier: Matrix Scientific

048084-500MG 048084-1G 048084-5G
101882-736EA 278.46 USD
101882-736 101882-738 101882-740
[2-(4-Methylpiperazino)-5-nitrophenyl]methanol ≥95%
[2-(4-Methylpiperazino)-5-nitrophenyl]methanol

Hazard: Irritant

Formula: C₁₂H₁₇N₃O₃
MDL Number: MFCD09972261
CAS Number: 802541-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR