2-Morpholino-5-nitrobenzaldehyde ≥95%

Supplier: Matrix Scientific

048077-500MG 048077-1G 048077-5G
101882-706EA 181.54 USD
101882-706 101882-708 101882-710
2-Morpholino-5-nitrobenzaldehyde ≥95%
2-Morpholino-5-nitrobenzaldehyde

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₄
MDL Number: MFCD00439921
CAS Number: 30742-62-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR