Ethyl-1-{[(2-bromoethyl)(tert-butoxycarbonyl)amino]sulfonyl}-3-piperidinecarboxylate ≥95%

Supplier: Matrix Scientific

048075-500MG 048075-1G
101882-698EA 375.38 USD
101882-698 101882-700
Ethyl-1-{[(2-bromoethyl)(tert-butoxycarbonyl)amino]sulfonyl}-3-piperidinecarboxylate ≥95%
Ethyl-1-{[(2-bromoethyl)(tert-butoxycarbonyl)amino]sulfonyl}-3-piperidinecarboxylate

Hazard: Irritant

Formula: C₁₅H₂₇BrN₂O₆S
MW: 443.37 g/mol
MDL Number: MFCD09972257
CAS Number: 1017782-89-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR