Ethyl-2-oxo-2-[4-(trifluoromethyl)piperidino]acetate ≥95%

Supplier: Matrix Scientific

048071-500MG 048071-1G
101882-680EA 556.92 USD
101882-680 101882-682
Ethyl-2-oxo-2-[4-(trifluoromethyl)piperidino]acetate ≥95%
Ethyl-2-oxo-2-[4-(trifluoromethyl)piperidino]acetate

Hazard: Irritant

Formula: C₁₀H₁₄F₃NO₃
MDL Number: MFCD09972256
CAS Number: 1017782-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR