Ethyl-2,5-dichloroisonicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2,5-dichloroisonicotinate, Ethyl 2,5-dichloropyridine-4-carboxylate

048064-1G 048064-5G
101882-644EA 113.85 USD
101882-644 101882-646
Ethyl-2,5-dichloroisonicotinate ≥95%
Ethyl 2,5-dichloroisonicotinate

Hazard: Irritant

Formula: C₈H₇Cl₂NO₂
MW: 220.0500 g/mol
MDL Number: MFCD09972252
CAS Number: 603122-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR