2-[4-(3-Nitro-2-pyridinyl)piperazino]-1-ethanol ≥95%
Supplier: Matrix Scientific
048059-500MG
048059-1G
048059-5G
101882-616EA
181.54
USD
101882-616
101882-618
101882-620
2-[4-(3-Nitro-2-pyridinyl)piperazino]-1-ethanol ≥95%
2-[4-(3-Nitro-2-pyridinyl)piperazino]-1-ethanol
Hazard: Irritant
Formula:
C₁₁H₁₆N₄O₃ |
MDL Number:
MFCD00278397 CAS Number: 215434-62-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...