2-Benzyl-6-oxa-2-azabicyclo[3.2.1]octan-7-one ≥95%

Supplier: Matrix Scientific

048035-500MG 048035-1G 048035-5G
101882-486EA 529.23 USD
101882-486 101882-488 101882-490
2-Benzyl-6-oxa-2-azabicyclo[3.2.1]octan-7-one ≥95%
2-Benzyl-6-oxa-2-azabicyclo[3.2.1]octan-7-one

Hazard: Irritant

Formula: C₁₃H₁₅NO₂
MDL Number: MFCD23701382
CAS Number: 133192-43-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR