1-[2-(Benzyloxy)ethyl]tetrahydro-4(1H)-pyridinone ≥95%

Supplier: Matrix Scientific

048022-500MG 048022-1G 048022-5G
101882-434EA 375.38 USD
101882-434 101882-436 101882-438
1-[2-(Benzyloxy)ethyl]tetrahydro-4(1H)-pyridinone ≥95%
1-[2-(Benzyloxy)ethyl]tetrahydro-4(1H)-pyridinone

Hazard: Irritant

Formula: C₁₄H₁₉NO₂
MDL Number: MFCD09152745
CAS Number: 948894-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR