1-{[Benzyl(tert-butoxycarbonyl)amino]sulfonyl}-3-piperidinecarboxylic acid ≥95%

Supplier: Matrix Scientific

047950-500MG 047950-1G 047950-5G
101882-120EA 375.38 USD
101882-120 101882-122 101882-124
1-{[Benzyl(tert-butoxycarbonyl)amino]sulfonyl}-3-piperidinecarboxylic acid ≥95%
1-{[Benzyl(tert-butoxycarbonyl)amino]sulfonyl}-3-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₁₈H₂₆N₂O₆S
MDL Number: MFCD09972238
CAS Number: 1017782-65-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR