Methyl-2-[4-(trifluoromethyl)piperidino]acetate ≥95%

Supplier: Matrix Scientific

047942-500MG 047942-1G
101882-092EA 346.15 USD
101882-092 101882-094
Methyl-2-[4-(trifluoromethyl)piperidino]acetate ≥95%
Methyl-2-[4-(trifluoromethyl)piperidino]acetate

Hazard: Irritant

Formula: C₉H₁₄F₃NO₂
MW: 225.21 g/mol
MDL Number: MFCD09865019
CAS Number: 860343-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR