2-(Benzyloxy)-1-ethanamine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(Benzyloxy)ethanamine, 2-(Benzyloxy)ethylamine

047920-1G 047920-5G 047920-25G
101882-004EA 44.62 USD
101882-004 101882-006 101882-008
2-(Benzyloxy)-1-ethanamine ≥95%
2-(Benzyloxy)-1-ethanamine

Hazard: Corrosive

Formula: C₉H₁₃NO
MW: 151.2056 g/mol
MDL Number: MFCD00235180
CAS Number: 38336-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR