5-(4-(2-(5-Ethylpyridin-2-yl)ethoxy)benzyl)thiazolidine-2,4-dione hydrochloride ≥99%
Supplier: Matrix Scientific
Synonyms:
Pioglitazone HCl, 5-[4-[2-(5-Ethyl-2-pyridyl)ethoxy]benzyl]-2,4-thiazolidinedione hydrochloride, Pioglitazone hydrochloride
047903-1G
047903-5G
101881-922EA
283.08
USD
101881-922
101881-924
5-(4-(2-(5-Ethylpyridin-2-yl)ethoxy)benzyl)thiazolidine-2,4-dione hydrochloride ≥99%
5-(4-(2-(5-Ethylpyridin-2-yl)ethoxy)benzyl)thiazolidine-2,4-dione hydrochloride
Hazard: Irritant
Formula:
C₁₉H₂₁ClN₂O₃S MW: 392.90 g/mol |
MDL Number:
MFCD04975446 CAS Number: 112529-15-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...