Azathioprine ≥95%

Supplier: Matrix Scientific

047900-500MG 047900-1G 047900-5G
101881-906EA 69.23 USD
101881-906 101881-908 101881-910
Azathioprine ≥95%
Azathioprine

Hazard: Irritant

Formula: C₉H₇N₇O₂S
MDL Number: MFCD00069203
CAS Number: 446-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR