Sertraline hydrochloride ≥98%

Supplier: Matrix Scientific

Synonyms: (1S,4S)-1-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-4-(methylamino)naphthalene Hydrochloride

047897-1G 047897-5G
101881-904EA 73.85 USD
101881-904 102663-730
Sertraline hydrochloride ≥98%
Sertraline hydrochloride

Hazard: Irritant

Formula: C₁₇H₁₇Cl₂N
MW: 342.69 g/mol
MDL Number: MFCD00895772
CAS Number: 79559-97-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR