Paroxetine hydrochloride hemihydrate ≥95%

Supplier: Matrix Scientific

Synonyms: (3S,4R)-(-)-3-[(1,3-Benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)piperidine Hydrochloride

047895-1G 047895-5G
101881-894EA 158.46 USD
101881-894 101881-896
Paroxetine hydrochloride hemihydrate ≥95%
Paroxetine hydrochloride hemihydrate

Hazard: Irritant

Formula: C₁₉H₂₀FNO₃
MW: 365.83 g/mol
MDL Number: MFCD03658863
CAS Number: 110429-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR