Etodolac ≥95%

Supplier: Matrix Scientific

Synonyms: Etodlic acid, 1,8-Diethyl-1,3,4,9-tetrahydropyrano[3,4-b]indole-1-acetic acid, 2-(1,8-Diethyl-1,3,4,9-tetrahydropyrano[3,4-b]indol-1-yl)acetic acid

047889-1G 047889-5G
101881-870EA 21.54 USD
101881-870 101881-872
Etodolac ≥95%
Etodolac

Hazard: Irritant

Formula: C₁₇H₂₁NO₃
MDL Number: MFCD00133313
CAS Number: 41340-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR