Clopidogrel bisulfate ≥98%

Supplier: Matrix Scientific

Synonyms: (±)-Clopidogrel hydrogen sulphate, Methyl 2-(2-chlorophenyl)-2-(6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl)acetate sulphate, (±)-Clopidogrel bisulphate

047886-1G 047886-5G
101881-856EA 246.15 USD
101881-856 102833-150
Clopidogrel bisulfate ≥98%
Clopidogrel bisulfate

Hazard: Irritant

Formula: C₁₆H₁₈ClNO₆S₂
MDL Number: MFCD05865229
CAS Number: 135046-48-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR