Cetirizine dihydrochloride ≥99%
Supplier: Matrix Scientific
Synonyms:
Cetirizine 2HCl, 2-(2-(4-((4-Chlorophenyl)(phenyl)methyl)piperazin-1-yl)ethoxy)acetic acid dihydrochloride
047885-5G
047885-25G
101881-850EA
121.54
USD
101881-850
101881-852
Cetirizine dihydrochloride ≥99%
Cetirizine dihydrochloride
Hazard: Irritant
Formula:
C₂₁H₂₅ClN₂O₃ MW: 461.81 g/mol |
MDL Number:
MFCD00941428 CAS Number: 83881-52-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...